missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Lysosomal alpha-Glucosidase/GAA, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 8329-GH-025
This item is not returnable.
View return policy
Description
Measured by its ability to release glucose from starch. The specific activity is >7,500 pmol/min/μg, as measured under the described conditions.Specifications
| P10253 | |
| 2548 | |
| Lysosomal alpha-Glucosidase | |
| 25 µg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 6 months, -20°C to -70°C under sterile conditions after opening. | |
| His | |
| <0.10 EU / 1 µg of the protein by the LAL method. | |
| Unconjugated |
| Supplied as a 0.2 µm filtered solution in Tris, NaCl and Glycerol. | |
| M.W. (Observed): 95-105 kDa, reducing conditions; M.W. (theoretical): 99 kDa | |
| SDS-PAGE: 95-105kDa, reducing conditions | |
| Human embryonic kidney cell, HEK293-derived human Lysosomal alpha-Glucosidase protein Ala70-Cys952, with an N-terminal 6-His tag | |
| RUO | |
| Carrier Protein | |
| GAA | |
| Yes |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?