missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Indoline-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 11466570
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466570 1 g 1g
15443737 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11466570 Supplier Thermo Scientific Alfa Aesar Supplier No. H27626.03
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98167-06-7
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00792496
InChI Key QNRXNRGSOJZINA-MRVPVSSYSA-N
Synonym r-indoline-2-carboxylic acid, r-+-indoline-2-carboxylic acid, 2r-2,3-dihydro-1h-indole-2-carboxylic acid, r---indoline-2-carboxylic acid, 2r-indoline-2-carboxylic acid, r-+-2,3-dihydroindole-2-carboxylic acid, r-2,3-dihydro-1h-indole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 2,3-dihydro-, 2r, pubchem20604
PubChem CID 6928266
IUPAC Name (2R)-2,3-dihydro-1H-indole-2-carboxylic acid
SMILES C1C(NC2=CC=CC=C21)C(=O)O

Specifications

Melting Point ∽177°C (decomposition)
Quantity 1 g
Formula Weight 163.18
Percent Purity 97%
Chemical Name or Material (R)-(+)-Indoline-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.