missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Mandelic acid, 99%

Product Code. 10417673 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10417673 5 g Glass bottle 5g
10001290 25 g Plastic bottle 25g
10376860 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10417673 Supplier Thermo Scientific Acros Supplier No. 125320050
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 611-71-2 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-71-2
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064251
InChI Key IWYDHOAUDWTVEP-SSDOTTSWSA-N
Synonym r-mandelic acid, r---mandelic acid, d-mandelic acid, r-2-hydroxy-2-phenylacetic acid, --mandelic acid, d--mandelic acid, d---mandelic acid, 2r-2-hydroxy-2-phenylacetic acid, r-alpha-hydroxyphenylacetic acid, d-2-phenylglycolic acid
PubChem CID 11914
ChEBI CHEBI:17656
IUPAC Name (2R)-2-hydroxy-2-phenylacetic acid
SMILES O[C@@H](C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 130°C to 135°C
CAS Min % 98.5
CAS Max % 100.0
Color Beige-Yellow to White
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder, Crystals or Flakes
Assay Percent Range 99%
Linear Formula C6H5CH(OH)CO2H
Formula Weight 152.15
Percent Purity ≥98.5%
Chemical Name or Material (R)-(-)-Mandelic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(-)-Mandelic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.