missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Mandelic acid, ChiPros 99+%, ee 99+%

Product Code. 11329655
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329655 5 g 5g
11339655 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329655 Supplier Thermo Scientific Alfa Aesar Supplier No. L19116.06
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 611-71-2 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-71-2
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064251
InChI Key IWYDHOAUDWTVEP-SSDOTTSWSA-N
Synonym r-mandelic acid, r---mandelic acid, d-mandelic acid, r-2-hydroxy-2-phenylacetic acid, #NAME?, d--mandelic acid, d---mandelic acid, 2r-2-hydroxy-2-phenylacetic acid, r-alpha-hydroxyphenylacetic acid, d-2-phenylglycolic acid
PubChem CID 11914
ChEBI CHEBI:17656
IUPAC Name (2R)-2-hydroxy-2-phenylacetic acid
SMILES O[C@@H](C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 131°C to 135°C
Beilstein 2691094
Formula Weight 152.15
Percent Purity ≥99%
Quantity 5 g
Optical Rotation -153° (c=5 in Water)
Sensitivity Light sensitive
Assay ee ≥99%
Chemical Name or Material (R)-(-)-Mandelic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.