missing translation for 'onlineSavingsMsg'
Learn More

(R)-Mertansine-13CD3, TRC

Product Code. 30471065
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30471065 1 mg 1mg
30478210 5 mg 5mg
30465130 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30471065 Supplier Toronto Research Chemicals Supplier No. TRCM2578011MG
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-Mertansine-13CD3
Molecular Formula 13C C34 D3 H45 Cl N3 O10 S
Quantity 1 mg
InChI Formula InChI=1S/C35H48ClN3O10S/c1-19-10-9-11-26(46-8)35(44)18-25(47-33(43)37-35)20(2)31-34(4,49-31)27(48-32(42)21(3)38(5)28(40)12-13-50)17-29(41)39(6)23-15-22(14-19)16-24(45-7)30(23)36/h9-11,15-16,20-21,25-27,31,44,50H,12-14,17-18H2,1-8H3,(H,37,43)/b11-9+,19-10+/t20-,21-,25+,26-,27+,31-,34+,35+/m1/s1/i5+1D3
SMILES [2H][13C]([2H])([2H])N([C@H](C)C(=O)O[C@H]1CC(=O)N(C)c2cc(C\C(=C\C=C\[C@@H](OC)[C@@]3(O)C[C@H](OC(=O)N3)[C@@H](C)[C@H]4O[C@@]14C)\C)cc(OC)c2Cl)C(=O)CCS
IUPAC Name [(1S,2R,3R,5S,6S,16E,18E,20R,21S)-11-chloro-21-hydroxy-12,20-dimethoxy-2,5,9,16-tetramethyl-8,23-dioxo-4,24-dioxa-9,22-diazatetracyclo[19.3.1.110,14.03,5]hexacosa-10(26),11,13,16,18-pentaen-6-yl] (2R)-2-[3-sulfanylpropanoyl(trideuterio(113C)methyl)amino]propanoate
Molecular Weight (g/mol) 742.299
Formula Weight 741.297
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.