missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ (R)-(-)-α-Methylhistamine dihydrobromide

Product Code. 18006052
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18006052 50 mg 50mg
30001101 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18006052 Supplier R&D Systems™ Supplier No. 0569/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, standard H3 agonist

Very potent, high affinity H3 agonist (KD = 50.3 nM) that displays > 200-fold selectivity over H4 receptors. Inhibits H3-mediated histamine synthesis and release in the CNS and stimulates H4-mediated eosinophil shape change (EC50 = 66 nM).

S-enantiomer also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868698-49-1
Molecular Formula C6H13Br2N3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 287
Purity >99%
Molecular Weight (g/mol) 286.999
Chemical Name or Material (R)-(-)-α-Methylhistamine dihydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.