Learn More
R&D Systems™ Mouse GLP-2R (NP_783612) VersaClone cDNA
Product Code. 18704418
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. Quantity Unit Size
18704418 10 μg 10µg
1 options
Product Code. 18704418 Supplier R&D Systems™ Supplier No. RDC1034
  • ONLINE EXCLUSIVE PRICE
    341.00 €
    322.35 € / 10µg
    Save 18.65 €
    5% Off
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
Glucagon-like Peptide 2 Receptor
The Mouse GLP-2R (NP_783612) VersaClone cDNA clone contains the complete ORF for GLP-2R, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The GLP-2R insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans