missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-N-(2-Hydroxyethyl)-α-phenylethylamine, 99%, Thermo Scientific™

Product Code. 10777202 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10777202 1 g Glass bottle 1g
10642912 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10777202 Supplier Thermo Scientific Acros Supplier No. 338820010
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 80548-31-8
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.23
MDL Number MFCD01862171
InChI Key GXIWMXAAPLZOBY-SECBINFHSA-N
Synonym r-+-2-1-phenylethyl amino ethanol, 2-1r-1-phenylethyl amino ethan-1-ol, 2-1r-1-phenylethyl amino ethanol, 2-r-alpha-methylbenzylamino ethanol, ethanol, 2-1r-1-phenylethyl amino, r-+-n-2-hydroxyethyl-phenylethylamine, r-+-n-2-hydroxyethyl-alpha-methylbenzylamine, r-+-n-2-hydroxyethyl-alpha-phenylethylamine
PubChem CID 6993821
IUPAC Name 2-[[(1R)-1-phenylethyl]amino]ethanol
SMILES CC(C1=CC=CC=C1)NCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5320 to 1.5350
Quantity 1 g
Specific Rotation Condition +44° (22°C c=1,C2H5OH)
Specific Rotation + 44.00
Formula Weight 165.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (R)-(+)-N-(2-Hydroxyethyl)-α-phenylethylamine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(+)-N-(2-Hydroxyethyl)-a-phenylethylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.