missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-N alpha-Dimethylbenzylamine, 99+%

Product Code. 10557624 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10557624 1 g Glass bottle 1g
10386624 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10557624 Supplier Thermo Scientific Acros Supplier No. 395850010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5933-40-4 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5933-40-4
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00044966
InChI Key RCSSHZGQHHEHPZ-MRVPVSSYSA-N
Synonym r-n-methyl-1-phenylethanamine, r-+-n,alpha-dimethylbenzylamine, r-n-methyl-alpha-phenylethylamine, methyl 1r-1-phenylethyl amine, r-+-n-methyl-1-phenylethylamine, unii-7iz3431sc3, r-+-n,a-dimethylbenzylamine, r-+-n,, a-dimethylbenzylamine, benzenemethanamine, n,alpha-dimethyl-, alphar, 1r-n-methyl-1-phenylethanamine
PubChem CID 2060888
IUPAC Name (1R)-N-methyl-1-phenylethanamine
SMILES CC(C1=CC=CC=C1)NC

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Colorless
Density 0.9240g/mL
Boiling Point 74.0°C to 76.0°C (11.0 mmHg)
Flash Point 61°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH(CH3)NHCH3
Refractive Index 1.5100 to 1.5120 (20°C, 589nm)
Quantity 1 g
Specific Gravity 0.924
Specific Rotation +63° to +65° (20°C, 589nm) (C=4, methanol)
Solubility Information Solubility in water: insoluble
Formula Weight 135.21
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material (R)-(+)-N α-Dimethylbenzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.