missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-BOC-4-Chlorophenylalanine, 95%, 98% ee, Thermo Scientific™

Product Code. 10457972 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10457972 1 g 1g
10733983 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10457972 Supplier Thermo Scientific Acros Supplier No. 362920010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 57292-44-1
Molecular Formula C14H18ClNO4
Molecular Weight (g/mol) 299.75
MDL Number MFCD00076978
InChI Key BETBOAZCLSJOBQ-LLVKDONJSA-N
Synonym boc-d-phe 4-cl-oh, boc-4-chloro-d-phenylalanine, boc-d-4-chlorophe, boc-d-4-chlorophenylalanine, n-tert-butoxycarbonyl-4-chloro-d-phenylalanine, n-boc-4-chloro-d-phenylalanine, r-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, r-n-boc-4-chlorophenylalanine, 2r-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, d-phenylalanine, 4-chloro-n-1,1-dimethylethoxy carbonyl
PubChem CID 7018791
IUPAC Name (2R)-3-(4-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CC1=CC=C(Cl)C=C1)C(O)=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 110°C
Color White
Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 299.75
Percent Purity 95%,98% ee
Physical Form Powder
Chemical Name or Material (R)-N-BOC-4-Chlorophenylalanine, 95%, (98% e.e.)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.