missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-BOC-alpha-Ethylalanine, 98%, 98% ee

Product Code. 11945521 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11945521 500 mg 500mg
11935521 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11945521 Supplier Thermo Scientific Acros Supplier No. 441065000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 123254-58-0 | C10H19NO4 | 217.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123254-58-0
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.27
MDL Number MFCD12963786
InChI Key SHZXLTCEPXVCSV-SNVBAGLBSA-N
Synonym boc-d-isovaline, r-2-tert-butoxycarbonyl amino-2-methylbutanoic acid, r-2-tert-butoxycarbonylamino-2-methylbutanoic acid, boc-iso-valine, r-n-boc-a-ethylalanine, s-n-boc-a-ethylalanine, r-2-methyl-2-tert-butoxycarbonylamino butyric acid, 2r-2-tert-butoxycarbonylamino-2-methyl-butanoic acid, 2r-2-tert-butoxy carbonyl amino-2-methylbutanoic acid, 2r-2-tert-butoxycarbonyl amino-2-methylbutanoic acid
PubChem CID 14284791
IUPAC Name (2R)-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CCC(C)(NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 104°C to 112°C
CAS Min % 97.5
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Specific Rotation -12
Quantity 500 mg
Formula Weight 217.27
Percent Purity 98%,98% ee
Physical Form Powder
Chemical Name or Material (R)-N-BOC-α-Ethylalanine, 98% ee
Show More Show Less
Product Identifier
  • (R)-N-BOC-alpha-Ethylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.