missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-BOC-piperidine-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10058049 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10058049 1 g Glass bottle 1g
10413530 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10058049 Supplier Thermo Scientific Acros Supplier No. 436500010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 163438-09-3
Molecular Formula C11H18NO4
Molecular Weight (g/mol) 228.27
MDL Number MFCD02179173
InChI Key NXILIHONWRXHFA-MRVPVSSYSA-M
Synonym r-boc-nipecotic acid, r-1-tert-butoxycarbonyl piperidine-3-carboxylic acid, r-n-boc-piperidine-3-carboxylic acid, boc-d-nip-oh, r-1-boc-piperidine-3-carboxylic acid, r-1-boc-nipecotic acid, d-1-boc-nipecotic acid, n-boc-r-nipecotic acid, r-boc-nip, r-boc-nip-oh
PubChem CID 643516
SMILES CC(C)(C)OC(=O)N1CCC[C@H](C1)C([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 166°C to 171°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation Condition −52.20° (20°C c=1.15 MeOH)
Specific Rotation -52.2
IUPAC Name (3R)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-3-carboxylic acid
Formula Weight 229.28
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (R)-N-BOC-Piperidine-3-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • (R)-N-BOC-Piperidine-3-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.