missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-BOC-Prolinal, 97+%

Product Code. 10493345 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10493345 5 g Glass Bottle 5g
10550934 1 g Glass Bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10493345

Brand: Thermo Scientific Acros 376230050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 73365-02-3 | C10H17NO3 | 199.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73365-02-3
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD01321389
InChI Key YDBPZCVWPFMBDH-MRVPVSSYSA-N
Synonym n-boc-d-prolinal, n-tert-butoxycarbonyl-d-prolinal, r-tert-butyl 2-formylpyrrolidine-1-carboxylate, boc-d-prolinal, tert-butyl 2r-2-formylpyrrolidine-1-carboxylate, 1-pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, 2r, tert-butyl r-2-formylpyrrolidine-1-carboxylate, r-n-boc-prolinal, pubchem15956
PubChem CID 7009153
IUPAC Name tert-butyl (2R)-2-formylpyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1C=O

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
NMR 97.0% min.
Assay Percent Range 97+%
Packaging Glass Bottle
Refractive Index 1.4600 to 1.4650 (20°C, 589nm)
Specific Rotation +90° to +102° (20°C, 589nm) (c=1, CHCl3)
Density 1.059g/mL
Specific Rotation Condition +91.50° (20°C C=1,chloroform)
Quantity 5 g
Formula Weight 199.25
Physical Form Liquid
Chemical Name or Material (R)-N-BOC-Prolinal
Show More Show Less
compliance-icons
Product Identifier
  • (R)-N-BOC-Prolinal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.