missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-BOC-Propargylglycine, 95%, 98% ee

Product Code. 10314172 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10314172 1 g Glass Bottle 1g
10157013 5 g Glass Bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10314172

Brand: Thermo Scientific Acros 362850010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 63039-46-3 | C10H14NO4 | 212.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63039-46-3
Molecular Formula C10H14NO4
Molecular Weight (g/mol) 212.23
MDL Number MFCD01320886
InChI Key AMKHAJIFPHJYMH-SSDOTTSWSA-M
Synonym boc-d-propargylglycine, r-2-tert-butoxycarbonyl amino pent-4-ynoic acid, n-boc-2-propargyl-d-glycine, boc-d-pra-oh, s-n-boc-propargylglycine, 2r-2-tert-butoxycarbonyl amino pent-4-ynoic acid, boc-d-gly propargyl-oh, boc-r-2-propargylglycine, n-tert-butoxycarbonyl-3-ethynyl-d-alanine, 2-r-tert-butoxycarbonylaminopent-4-ynoic acid
PubChem CID 7009126
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-ynoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CC#C)C([O-])=O

Specifications

Melting Point 93.0°C to 98.0°C
CAS Min % 94.0
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 213.23
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material (R)-N-BOC-Propargylglycine, 98% ee
Show More Show Less
compliance-icons
Product Identifier
  • (R)-N-BOC-Propargylglycine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.