missing translation for 'onlineSavingsMsg'
Learn More

R-(+)-Nicotine, TRC

Product Code. 30133806
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133806 50 mg 50mg
30133567 25 mg 25mg
17907669 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133806 Supplier Toronto Research Chemicals Supplier No. TRCN41243050MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25162-00-9
Molecular Formula C10 H14 N2
Molecular Weight (g/mol) 162.23
Synonym 3-[(2R)-1-Methyl-2-pyrrolidinyl]pyridine, 2'βH-Nicotine, (R)-3-(1-Methyl-2-pyrrolidinyl)pyridine, (+)-(R)-Nicotine, (+)-Nicotine, (R)-(+)-Nicotine, (R)-3-(1-Methylpyrrolidin-2-yl)pyridine, (R)-Nicotine, D-(+)-Nicotine, D-Nicotine, Pseudonicotine, d-Nicotine
IUPAC Name 3-[(2R)-1-methylpyrrolidin-2-yl]pyridine
SMILES CN1CCC[C@@H]1c2cccnc2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H14N2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9/h2,4,6,8,10H,3,5,7H2,1H3/t10-/m1/s1
Formula Weight 162.1157
Chemical Name or Material (+)-Nicotine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.