missing translation for 'onlineSavingsMsg'
Learn More

(R)-Norfluoxetine Phthalimide, TRC

Product Code. 30427108
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427108 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Artikelnummer. 30427108 Leverandør Toronto Research Chemicals Leverandørnr. TRCN6811405MG
 Restricted Item
Estimated Shipment: 10-12-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material (R)-Norfluoxetine Phthalimide
CAS 878663-12-8
Molecular Formula C24H18F3NO3
Quantity 5 mg
Synonym 2-[(3R)-3-Phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]-1H-isoindole-1,3(2H)-dione, (R)-1-(Phthalimido)-3-[(4-trifluoromethylphenyl)oxy]-3-phenylpropane
InChI Formula InChI=1S/C24H18F3NO3/c25-24(26,27)17-10-12-18(13-11-17)31-21(16-6-2-1-3-7-16)14-15-28-22(29)19-8-4-5-9-20(19)23(28)30/h1-13,21H,14-15H2/t21-/m1/s1
SMILES FC(F)(F)c1ccc(O[C@H](CCN2C(=O)c3ccccc3C2=O)c4ccccc4)cc1
IUPAC Name 2-[(3R)-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]isoindole-1,3-dione
Molecular Weight (g/mol) 425.4
Formula Weight 425.1239
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.