missing translation for 'onlineSavingsMsg'
Learn More

(R)-Omeprazole Sodium Salt, TRC

Product Code. 30007969
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007969 1 mg 1mg
30006725 2.5 mg 2.5mg
30006962 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007969 Supplier Toronto Research Chemicals Supplier No. TRCO6350101MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161796-77-6
Molecular Formula C17 H18 N3 O3 S . Na
Molecular Weight (g/mol) 367.4
Synonym 6-Methoxy-2-[(R)-[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl]-1H-benzimidazole sodium salt (1:1), 1H-Benzimidazole, 5-methoxy-2-[(R)-[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl]-, sodium salt (9CI), 1H-Benzimidazole, 5-methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl]-, sodium salt, (R)-, (R)-Omeprazole sodium
IUPAC Name sodium;5-methoxy-2-[(R)-(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]benzimidazol-1-ide
SMILES [Na+].COc1ccc2[n-]c(nc2c1)[S@](=O)Cc3ncc(C)c(OC)c3C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C17H18N3O3S.Na/c1-10-8-18-15(11(2)16(10)23-4)9-24(21)17-19-13-6-5-12(22-3)7-14(13)20-17;/h5-8H,9H2,1-4H3;/q-1;+1/t24-;/m1./s1
Formula Weight 367.0967
Chemical Name or Material (R)-Omeprazole sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.