missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ (R)-(-)-Phenylephrine hydrochloride

Product Code. 18712474
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18712474 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18712474 Supplier R&D Systems™ Supplier No. 2838/100
  • ONLINE EXCLUSIVE PRICE
    156.54 €
    138.60 € / 100mg
    Save 17.94 €
    11% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. α1 agonist

α1-adrenoceptor agonist; pKi values are 5.86, 4.87 and 4.70 for α1D, α1B and α1A receptors respectively.
TRUSTED_SUSTAINABILITY

Specifications

CAS 61-76-7
Quantity 100 mg
Product Line Tocris
Molecular Formula C9H14ClNO2
Formula Weight 203.67
Purity >99%
Molecular Weight (g/mol) 203.67
Chemical Name or Material (R)-(-)-Phenylephrine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.