missing translation for 'onlineSavingsMsg'
Learn More

(R)-alpha-Propargylglycine, 98%, 98% ee

Product Code. 11945371 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11945371 100 mg 100mg
11955371 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11945371 Supplier Thermo Scientific Acros Supplier No. 441221000
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 23235-03-2 | C5H7NO2 | 113.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23235-03-2
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.12
InChI Key DGYHPLMPMRKMPD-SCSAIBSYSA-N
Synonym d-propargylglycine, r-2-aminopent-4-ynoic acid, r-alpha-propargylglycine, unii-2252qef2pl, 4-pentynoic acid, 2-amino-, 2r, 2r-2-aminopent-4-ynoic acid, r-2-propargylglycine, r-2-amino-4-pentynoic acid, d-2-propynylglycine, propargylglycine, d
PubChem CID 6992713
IUPAC Name (2R)-2-aminopent-4-ynoic acid
SMILES C#CCC(C(=O)O)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >180°C
Color White to Yellow
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5%
Packaging Glass bottle
Quantity 100 mg
Specific Rotation 31.65
Formula Weight 113.12
Percent Purity 98%,98% ee
Physical Form Powder
Chemical Name or Material (R)-α-Propargylglycine, 98% ee
Show More Show Less
compliance-icons
Product Identifier
  • (R)-alpha-Propargylglycine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.