missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Propylene oxide, 98+%

Product Code. 10396594 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10396594 5 g Glass bottle 5g
10193104 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10396594 Supplier Thermo Scientific Acros Supplier No. 399570050
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15448-47-2 | C3H6O | 58.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15448-47-2
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00066211
InChI Key GOOHAUXETOMSMM-GSVOUGTGSA-N
Synonym r-+-propylene oxide, r-propylene oxide, 2r-2-methyloxirane, r-2-methyloxirane, r-methyloxirane, +-propylene oxide, r-epoxypropane, +-methyloxirane, r-+-1,2-epoxypropane, r-1,2-epoxypropane
PubChem CID 146261
ChEBI CHEBI:28985
IUPAC Name (2R)-2-methyloxirane
SMILES CC1CO1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -112.0°C
Density 0.8290g/mL
Boiling Point 33.0°C to 34.0°C
Flash Point −37°C
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 0.829
Solubility Information Solubility in water: 405g/L (20°C)
Viscosity 0.38 mPa.s (10°C)
Formula Weight 58.08
Percent Purity 98+%
Chemical Name or Material (R)-(+)-Propylene oxide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Identificativo prodotto
  • (R)-(+)-Propylene oxide
Avvertenza
  • Danger
Categoria di pericolo
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 1
  • Germ cell mutagenicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H224-Extremely flammable liquid and vapour.
Consigli di prudenza
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P403+P235-Store in a well-ventilated place. Keep cool.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.