missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Propylene oxide, 99%

Product Code. 11434947
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434947 5 g 5g
11444947 25 g 25g
11454947 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434947 Supplier Thermo Scientific Alfa Aesar Supplier No. B21884.06
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15448-47-2 | C3H6O | 58.08 g/mol

(R)-(+)-Propylene oxide is widely utilized in the manufacture of polyether polyols and propylene glycol, which is used to make the polyurethane plastics. It finds application as a fumigant for the sterilization of packaged foods and plastic medical instruments. Combine with ethanol, it used to make biological samples for electron microscopy. It also used in model aircraft and surface vehicles as a glow fuel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-Propylene oxide is widely utilized in the manufacture of polyether polyols and propylene glycol, which is used to make the polyurethane plastics. It finds application as a fumigant for the sterilization of packaged foods and plastic medical instruments. Combine with ethanol, it used to make biological samples for electron microscopy. It also used in model aircraft and surface vehicles as a glow fuel.

Solubility
Miscible with water and many organic solvents.

Notes
Highly flammable. Keep away from sources of ignition. The substance may polymerize under the influence of bases, acids and metal chlorides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15448-47-2
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00066211
InChI Key GOOHAUXETOMSMM-GSVOUGTGSA-N
Synonym r-+-propylene oxide, r-propylene oxide, 2r-2-methyloxirane, r-2-methyloxirane, r-methyloxirane, +-propylene oxide, r-epoxypropane, +-methyloxirane, r-+-1,2-epoxypropane, r-1,2-epoxypropane
PubChem CID 146261
ChEBI CHEBI:28985
IUPAC Name (2R)-2-methyloxirane
SMILES CC1CO1

Specifications

Density 0.829
Boiling Point 34°C to 35°C
Flash Point −37°C (−35°F)
Odor Ether-like
Quantity 5 g
UN Number UN1280
Beilstein 79764
Merck Index 14,7856
Solubility Information Miscible with water and many organic solvents.
Formula Weight 58.08
Percent Purity 99%
Chemical Name or Material (R)-(+)-Propylene oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.