missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Quantikine Immunoassay Control Set 934 for Human IL-15

Product Code. 30001857
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001857 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001857 Supplier R&D Systems™ Supplier No. QC26

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use with catalog numbers D1500, S1500, PD1500

For use as quantitative controls for the determination of cytokine concentrations in biological fluids. Concentrations have been assigned using R&D Systems’ Quantikine kits. Controls are prepared in diluted porcine serum with preservatives. They contain recombinant human cytokines at low, medium and high concentrations. Controls are supplied lyophilized.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Detectable Analytes IL-15
Gene ID (Entrez) 3600
Intended Use Control
Quantity 1 Each
Source Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.