missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Quantikine Immunoassay Control Set 787 for Human TFF3
Beskrivelse
Quantikine Immunoassay Control Set 787 is designed for use as quantitative controls for the determination of human TFF3 concentrations in biological fluids.
- Analyte: Recombinant human TFF3
- Format: Lyophilized
- Composition: Diluted porcine serum with preservatives
- Intended Use: Quantitative controls for use with R and D System™ Quantikine™ kits
- Storage: Store unopened product at 2°C to 8°C
- Usage: Reconstitute with deionized or distilled water as indicated on the product datasheet
Tekniske data
Tekniske data
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Detectable Analytes | TFF3 |
| Sample Type | Biological Fluids |
| Type | Immunoassay Control |
| Gene ID (Entrez) | 7033 |
| Intended Use | Control |
| Product Line | R&D Systems |
| Quantity | 1 Each |
| Source | Human |
| Form | Lyophilized |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?