missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-Palonosetron Hydrochloride, TRC

Product Code. 30134038
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134038 10 mg 10mg
17949354 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134038 Supplier Toronto Research Chemicals Supplier No. TRCP16580510MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135729-75-8
Molecular Formula C19 H24 N2 O . HCl
Molecular Weight (g/mol) 332.87
Synonym 1H-Benz[de]isoquinolin-1-one, 2-(3R)-1-azabicyclo[2.2.2]oct-3-yl-2,3,3a,4,5,6-hexahydro-, hydrochloride (1:1), (3aR)-, 1H-Benz[de]isoquinolin-1-one, 2-(1-azabicyclo[2.2.2]oct-3-yl)-2,3,3a,4,5,6-hexahydro-, monohydrochloride, [R-(R*,R*)]- (9CI), (3aR)-2-(3R)-1-Azabicyclo[2.2.2]oct-3-yl-2,3,3a,4,5,6-hexahydro-1H-benz[de]isoquinolin-1-one hydrochloride (1:1), RS 25259-198
IUPAC Name (3aR)-2-[(3R)-1-azabicyclo[2.2.2]octan-3-yl]-3a,4,5,6-tetrahydro-3H-benzo[de]isoquinolin-1-one;hydrochloride
SMILES Cl.O=C1N(C[C@@H]2CCCc3cccc1c23)[C@H]4CN5CCC4CC5

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H24N2O.ClH/c22-19-16-6-2-4-14-3-1-5-15(18(14)16)11-21(19)17-12-20-9-7-13(17)8-10-20;/h2,4,6,13,15,17H,1,3,5,7-12H2;1H/t15-,17-;/m0./s1
Formula Weight 332.1655
Chemical Name or Material (R,R)-Palonosetron Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.