missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-Solifenacin Succinate, TRC

Product Code. 30426268
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426268 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426268 Supplier Toronto Research Chemicals Supplier No. TRCS676690100MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R,R)-Solifenacin Succinate
CAS 862207-70-3
Molecular Formula C27H32N2O6
Quantity 100 mg
Synonym (1R)-(3R)-1-Azabicyclo[2.2.2]oct-3-yl 3,4-Dihydro-1-phenyl-2(1H)- isoquinolinecarboxylate Butanedioic Acid, (1R,3'R)-Solifenacin Succinate, Rasageline Allyl Impurity
InChI Formula InChI=1S/C23H26N2O2.C4H6O4/c26-23(27-21-16-24-13-10-18(21)11-14-24)25-15-12-17-6-4-5-9-20(17)22(25)19-7-2-1-3-8-19;5-3(6)1-2-4(7)8/h1-9,18,21-22H,10-16H2;1-2H2,(H,5,6)(H,7,8)/t21-,22+;/m0./s1
SMILES C1CN2CCC1[C@H](C2)OC(=O)N3CCC4=CC=CC=C4C3C5=CC=CC=C5.C(CC(=O)O)C(=O)O
IUPAC Name [(3R)-1-azabicyclo[2.2.2]octan-3-yl] (1R)-1-phenyl-3,4-dihydro-1H-isoquinoline-2-carboxylate;butanedioic acid
Molecular Weight (g/mol) 480.55
Formula Weight 480.226
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.