missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human GDF-9 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in HCl.
Unit Size:
10µg
Description
The Recombinant Human GDF-9 Protein is derived from HEK293. The Recombinant Human GDF-9 Protein has been validated for the following applications: Bioactivity.
Order Info
The product is shipped at ambient temperature. Upon receipt, store it immediately
Specifications
Specifications
| Accession Number | O60383 |
| Formulation | Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. |
| Molecular Weight (g/mol) | 16 kDa (monomer) |
| Name | GDF-9 |
| Quantity | 10 μg |
| Source | Human embryonic kidney cell,HEK293-derived human GDF-9 protein Gly320-Arg454 (Gly391Arg) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | >95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction