missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (R)-(-)-Rolipram
GREENER_CHOICE

Product Code. 30001239
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001239

Brand: Tocris Bioscience™ 1349/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PDE4 inhibitor. More active enantiomer of rolipram (Cat. No. 0905)

More active enantiomer of the PDE4 inhibitor rolipram; 2-10-fold more potent than the S-(+) enantiomer.

Racemate and S-enantiomer also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85416-75-7
Molecular Formula C16H21NO3

Specifications

Quantity 10 mg
Formula Weight 275.35
Purity >98%
Molecular Weight (g/mol) 275.348
Chemical Name or Material (R)-(-)-Rolipram
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.