missing translation for 'onlineSavingsMsg'
Learn More

(R,S)-2,2-Dimethyl-1,3-dioxolane-4-methanol, TRC

Product Code. 30012307
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012307 50 g 50g
30008707 250 g 250g
30008810 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012307 Supplier Toronto Research Chemicals Supplier No. TRCD47264050G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-79-8
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
Synonym (RS)-Solketal, (+/-)-1,2-O-Isopropylideneglycerol, (+/-)-Glycerol 1,2-Acetonide, 1,2-Isopropylideneglycerin, 1,2-Isopropylideneglycerol, 2,3-(Isopropylidenedioxy)propanol, 2,3-Isopropylideneglycerol, Acetone Glyceryl Acetal, Dioxolan, α,β-Isopropylidene Ether, Glycerolacetone, NSC 59720;
SMILES CC1(C)OCC(CO)O1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C6H12O3/c1-6(2)8-4-5(3-7)9-6/h5,7H,3-4H2,1-2H3
Formula Weight 132.0786
Chemical Name or Material (R,S)-2,2-Dimethyl-1,3-dioxolane-4-methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.