missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-S-Methyl-S-phenylsulfoximine, 97%

Product Code. 11342586
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342586 250 mg 250mg
11332586 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342586 Supplier Thermo Scientific Alfa Aesar Supplier No. L08562.MD
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 60933-65-5 | C7H9NOS | 155.215 g/mol

(R)-(-)-S-Methyl-S-phenylsulfoximine is used as chiral ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-S-Methyl-S-phenylsulfoximine is used as chiral ligands.

Notes
Hygroscopic. Store away from water and oxidizing agents. Protect from humidity and water. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60933-65-5
Molecular Formula C7H9NOS
Molecular Weight (g/mol) 155.215
MDL Number MFCD00151460
InChI Key YFYIDTVGWCYSEO-JTQLQIEISA-N
Synonym s-+-s-methyl-s-phenylsulfoximine, s-+-s-methyl-s-phenylsulfoximine ee, s-imino methyl phenyl-??-sulfanone, r---s-methyl-s-phenylsulfoximine ee, imino-methyl-oxo-phenyl-$l^ 6, s-+-s-methyl-s-phenylsulphoximine, sulfoximine,s-methyl-s-phenyl-, s s
PubChem CID 25036288
IUPAC Name imino-methyl-oxo-phenyl-$l^{6}-sulfane
SMILES CS(=N)(=O)C1=CC=CC=C1

Specifications

Melting Point 29°C to 31°C
Quantity 250 mg
Beilstein 5245635
Sensitivity Hygroscopic
Optical Rotation −36° (c=1 in Acetone)
Formula Weight 155.22
Percent Purity 97%
Chemical Name or Material (R)-(-)-S-Methyl-S-phenylsulfoximine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.