missing translation for 'onlineSavingsMsg'
Learn More

(R,S)-N-Ethyl Nornicotine, TRC

Product Code. 30384710
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384710 5 mg 5mg
30374959 10 mg -
30377327 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384710 Supplier Toronto Research Chemicals Supplier No. TRCE9253005MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86900-39-2
Molecular Formula C11 H16 N2
Molecular Weight (g/mol) 176.26
Synonym Pyridine, 3-(1-ethyl-2-pyrrolidinyl)-, Pyridine, 3-(1-ethyl-2-pyrrolidinyl)-, (±)-, 3-(1-Ethyl-2-pyrrolidinyl)pyridine, Homonicotine, (±)-N-Ethylnornicotine, rac-Ethylnornicotine, rac-N-Ethylnornicotine
IUPAC Name 3-(1-ethylpyrrolidin-2-yl)pyridine
SMILES CCN1CCCC1c2cccnc2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C11H16N2/c1-2-13-8-4-6-11(13)10-5-3-7-12-9-10/h3,5,7,9,11H,2,4,6,8H2,1H3
Formula Weight 176.1313
Chemical Name or Material (R,S)-N-Ethylnornicotine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.