missing translation for 'onlineSavingsMsg'
Learn More

(R,S,S)-Enalapril Maleate, TRC

Product Code. 30135490
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135490 50 mg 50mg
30010100 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135490 Supplier Toronto Research Chemicals Supplier No. TRCE55524550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1356932-13-2
Synonym N-[(1R)-1-(Ethoxycarbonyl)-3-phenylpropyl]-L-alanyl-L-proline maleate∣(R)-1-[N-[1-(Ethoxycarbonyl)-3-phenylpropyl]-L-alanyl]-L-proline maleate∣(2S)-1-[(2S)-2-[[(1R)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]propanoyl]pyrrolidine-2-carboxylic acid maleate∣(R,S,S)-Enalapril maleate

Specifications

Format Neat
Molecular Formula C24H32N2O9
Molecular Weight (g/mol) 492.52, 492.53
InChI Key OYFJQPXVCSSHAI-BMSLDGIRSA-N
SMILES OC(=O)\C=C/C(O)=O.CCOC(=O)[C@@H](CCC1=CC=CC=C1)N[C@@H](C)C(=O)N1CCC[C@H]1C(O)=O
IUPAC Name (2S)-1-[(2S)-2-{[(2R)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoyl]pyrrolidine-2-carboxylic acid; (2Z)-but-2-enedioic acid
Product Type Impurity
Quantity 50 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.