missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-sec-Butanol, 99%

Product Code. 10296840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10296840 1 g Glass Bottle 1g
10781871 5 g Glass Bottle 5g
10762485 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10296840 Supplier Thermo Scientific Acros Supplier No. 217230010
  • ONLINE EXCLUSIVE PRICE
    61.40 €
    59.60 € / 1g
    Save 1.80 €
    2% Off
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 14898-79-4 | C4H10O | 74.123 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14898-79-4
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00064280
InChI Key BTANRVKWQNVYAZ-SCSAIBSYSA-N
Synonym r---2-butanol, r-2-butanol, --2-butanol, 2r-butan-2-ol, r-butan-2-ol, r---sec-butanol, 2-butanol, 2r, unii-dlh38k423j, r---sec-butyl alcohol
PubChem CID 84682
ChEBI CHEBI:35686
IUPAC Name (2R)-butan-2-ol
SMILES CCC(C)O

Specifications

CAS Min % 98.5
Color Colorless
CAS Max % 100.0
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3960 to 1.3980 (20°C, 589nm)
Assay Percent Range 99%
Linear Formula C2H5CH(OH)CH3
Beilstein 01, I, 189
Merck Index 15, 1543
Formula Weight 74.12
Percent Purity ≥98.5%
Chemical Name or Material (R)-(-)-sec-Butanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (R)-(-)-sec-Butanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.