missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Tetrahydrofurfuryl alcohol, 98+%, Thermo Scientific™

Product Code. 11339585
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11339585 250 mg 250mg
11329585 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339585 Supplier Thermo Scientific Alfa Aesar Supplier No. L19044.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22415-59-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD03093085
InChI Key BSYVTEYKTMYBMK-UHFFFAOYNA-N
Synonym r-tetrahydrofurfuryl alcohol, r-tetrahydrofuran-2-yl methanol, 2-furanmethanol, tetrahydro-, 2r, 2r-oxolan-2-yl methanol, r---tetrahydrofurfuryl alcohol, 2r-oxolan-2-ylmethanol, r---tetrahydrofurfurylalcohol, 2r-tetrahydrofuran-2-yl methanol, 2r-2-oxolanyl methanol, r-tetrahydrofuran-2-methanol
PubChem CID 6931745
IUPAC Name (oxolan-2-yl)methanol
SMILES OCC1CCCO1

Specifications

Density 1.053
Boiling Point 173°C to 175°C
Flash Point 75°C (167°F)
Refractive Index 1.452
Quantity 250 mg
Beilstein 79846
Optical Rotation −2.3° (Neat)
Formula Weight 102.13
Percent Purity ≥98%
Chemical Name or Material (R)-(-)-Tetrahydrofurfuryl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.