missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Tetrahydrofurfurylamine, 98+%, Thermo Scientific™

Product Code. 10252942 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10252942 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10252942 Supplier Thermo Scientific Acros Supplier No. 352630010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 7202-43-9
CAS Min % 98.0
Color Colorless
CAS Max % 100.0
Density 0.9800g/mL
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4540 to 1.456
Assay Percent Range 98% min. (GC)
Specific Gravity 0.98
Molecular Formula C5H11NO
MDL Number MFCD00192476
Synonym r---tetrahydrofurfurylamine, r-tetrahydrofuran-2-yl methanamine, r-tetrahydrofurfurylamine, 2r-oxolan-2-ylmethanamine, 2r-tetrahydro-2-furanmethanamine, 2-furanmethanamine, tetrahydro-, 2r, 1-2r-tetrahydrofuran-2-yl methanamine, pubchem7001, 2r-tetrahydrofurfurylamine
Boiling Point 151°C to 152°C
Solubility Information Solubility in water: miscible with water
InChI Key YNOGYQAEJGADFJ-RXMQYKEDSA-N
SMILES C1CC(OC1)CN
IUPAC Name [(2R)-oxolan-2-yl]methanamine
Molecular Weight (g/mol) 101.15
PubChem CID 2734124
Formula Weight 101.15
Percent Purity 98+%
Chemical Name or Material (R)-(-)-Tetrahydrofurfurylamine, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.