missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Tetrahydrofurfurylamine, 98+%

Product Code. 11363516
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363516 250 mg 250mg
11353516 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363516 Supplier Thermo Scientific Alfa Aesar Supplier No. L14410.MD
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 7202-43-9 | C5H11NO | 101.149 g/mol

(R)-(-)-Tetrahydrofurfurylamine is used in the synthesis of novel topoisomerase I targeting anti-cancer agents with mild to potent cytotoxic activity. Also used in the discovery and synthesis of orally bioavailable stearoyl-CoA desaturase 1 inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-Tetrahydrofurfurylamine is used in the synthesis of novel topoisomerase I targeting anti-cancer agents with mild to potent cytotoxic activity. Also used in the discovery and synthesis of orally bioavailable stearoyl-CoA desaturase 1 inhibitors.

Solubility
Fully miscible with water.

Notes
Hygroscopic, store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7202-43-9
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00192476
InChI Key YNOGYQAEJGADFJ-RXMQYKEDSA-N
Synonym r---tetrahydrofurfurylamine, r-tetrahydrofuran-2-yl methanamine, r-tetrahydrofurfurylamine, 2r-oxolan-2-ylmethanamine, 2r-tetrahydro-2-furanmethanamine, 2-furanmethanamine, tetrahydro-, 2r, 1-2r-tetrahydrofuran-2-yl methanamine, pubchem7001, 2r-tetrahydrofurfurylamine
PubChem CID 2734124
IUPAC Name [(2R)-oxolan-2-yl]methanamine
SMILES C1CC(OC1)CN

Specifications

Density 0.98
Quantity 250 mg
Refractive Index 1.455
UN Number UN2943
Beilstein 4652873
Boiling Point 152°C to 153°C
Solubility Information Fully miscible with water.
Formula Weight 101.15
Percent Purity ≥98%
Chemical Name or Material (R)-(-)-Tetrahydrofurfurylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.