missing translation for 'onlineSavingsMsg'
Learn More

Brady™ R10000 Series Ribbon for M7 Printers

Product Code. 17878764
Change view
Click to view available options
Color:
Blue
Green
White
Yellow
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17878764 Green Each
17808804 White Each
17891489 Blue Each
17838764 Yellow Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17878764 Supplier Brady™ Supplier No. 173197
Add to basket
Add to basket
This item is not returnable. View return policy

Featuring a 5-second 'drop and click' supply change, this smear-proof M7 printer ribbon needs no threading or calibration. Its super-tough construction makes it ideal for outdoor and industrial use.

  • Ideal for marking pipes and valves, facility safety and compliance ID and general ID
  • Offers strong solvent and scratch resistance, plus high-performance images
  • Self-contained M7 printer ribbon cartridges insert straight into the printer with ease

Specifications

Product Type Printer Ribbon
Color Green
For Use With (Equipment) Brady M710 Printer, BMP71 Label Printer
Material Resin
Length (English) 150 ft.
Width (English) 1 in.
Width (Metric) 5 cm
Dimensions (L x W) 150 ft. x 2 in.
Autoclavable Not Autoclavable
Sterility Non-sterile
Format Cartridge, Ribbon
Alternate Part Number M7-R10000-GN
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.