Learn More
Gilson™ Minipuls™ 3 R2/HF High-Flow Pump Head
Product Code. 10327522 Shop All Gilson Products
Change view
Click to view available options
Flow Rate:
1000μL/min. to 220 mL/min.
Unit Size:
Each
Product Code. Flow Rate Unit Size
10327522 1000μL/min. to 220 mL/min. Each
1 options
Product Code. 10327522 Supplier Gilson™ Supplier No. F117830
 Restricted Item
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with Peristaltic pump
  • High flow model
  • With 5 stainless steel rollers per head
  • Two channels
TRUSTED_SUSTAINABILITY
Specifications
Flow Rate 1000μL/min. to 220 mL/min.
Model R2/HF High-Flow Pump Head
No. of Channels 2
No. of Rollers 5
Roller Material Stainless Steel
Type Peristaltic Pump Head