Learn More
Melford R2A Agar
Description
Low nutrient medium for plate detection of heterotropic bacteria in water.
Low nutrient medium for plate detection of heterotropic bacteria in water. Low concentrations of carbon and energy sources enhance the recovery of stressed bacteria.
Formula per liter
Casein Digest Peptone: 0.25g
Peptic Digest of Animal Tissue: 0.25g
Magnesium Sulfate Heptahydrate: 0.05g
Sodium Pyruvate: 0.3g
Casein Acid Hydrolysate: 0.5g
Dipotassium Phosphate: 0.3g
Yeast Extract: 0.5g
Soluble Starch: 0.5g
Agar: 15.0g
Glucose: 0.5g
Specifications
Specifications
| Quantity | 1 kg |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.