Learn More
Epredia™ RA Lamb Biopsy Inserts, Biopsy insert pad, white
Product Code. 12628366 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Insert Pad
Unit Size:
Pack of 100
Product Code. Product Type Unit Size
12628366 Insert Pad Pack of 100
1 options
Product Code. 12628366 Supplier Epredia™ Supplier No. E10/87/B
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Prevent the loss of small or friable material while allowing interchange of fluids with Epredia™ RA Lamb Biopsy Inserts.
  • Mesh fits inside processing cassettes
  • Mesh hole size is 0.5mm
TRUSTED_SUSTAINABILITY
Specifications
Product Type Insert Pad
Material Stainless Steel
Color Blue