missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Canine IgG (H+L) Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-60673
559.30 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Canine samples. It is validated for ELISA.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| Rabbit | |
| Affinity Purified | |
| Secondary | |
| Canine | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 7.56mg/mL | |
| Western Blot 1:500-1:2500, ELISA 1:2000-1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin | |
| IGHG1 | |
| Dog IgG whole molecule | |
| 10 mg | |
| This product was prepared from monospecific antiserum by immunoaffinity chromatography using Dog IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction