Learn More
Rabbit anti-Cat IgM mu heavy chain,Polyclonal Secondary Antibody, Abnova™
Brand:  Abnova PAB9161.50mg
Product Code. 16264610
  • 979.00 € / 50mg
Specifications
Rabbit anti-Cat IgM mu heavy chain
Polyclonal
Rabbit anti-cat IgG recognizes cat IgM mu heavy chain. This secondary antibody was purified using a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above.
Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Cat IgM mu heavy chain
50 mg
No reaction was observed against cat IgG.
Store at 4°C on dry atmosphere. After reconstitution with 5mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
ELISA, Immunohistochemistry, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
Rabbit
This product is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above.
Secondary
Feline
Lyophilized