missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Dog IgM mu heavy chain,Polyclonal Secondary Antibody, Abnova™
Brand: Abnova PAB9186.50mg
This item is not returnable.
View return policy
Specifications
| Rabbit anti-Dog IgM mu heavy chain | |
| Polyclonal | |
| Rabbit anti-dog IgG recognizes dog IgM mu heavy chain. This secondary antibody was purified using a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. | |
| Lyophilized | |
| Dog IgM mu heavy chain | |
| 50 mg | |
| No reaction was observed against dog IgG. | |
| Store at 4°C on dry atmosphere. After reconstitution with 5mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| This product is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. | |
| Secondary | |
| Dog | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction