Learn More
Rabbit anti-Goat IgM mu heavy chain, Polyclonal, Secondary Antibody, Abnova™
Brand:  Abnova PAB9223.50mg
Product Code. 16031528
  • 979.00 € / 50mg
Specifications
Rabbit anti-Goat IgM mu heavy chain
Polyclonal
Rabbit anti-goat IgG recognizes goat IgM mu heavy chain. This secondary antibody was purified using a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above.
Liquid
Goat IgM mu heavy chain
50 mg
Goat
Liquid
ELISA, Immunohistochemistry, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
Rabbit
This product is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above.
Secondary
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.