missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Hamster IgG (H+L) Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
107.00 € - 215.00 €
Specifications
| Antigen | IgG (H+L) |
|---|---|
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoassay, Immunomicroscopy 1:10 - 1:500, Chemiluminescence Immunoassay (CIA) (CIA) |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18563221
|
Novus Biologicals
NBP1-74888-0.5mg |
0.5 mg |
107.00 €
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18507692
|
Novus Biologicals
NBP1-74888 |
2 mg |
215.00 €
2mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Hamster samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence ImmunoassaySpecifications
| IgG (H+L) | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Immunology | |
| 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoassay, Immunomicroscopy 1:10 - 1:500, Chemiluminescence Immunoassay (CIA) (CIA) | |
| Polyclonal | |
| Purified | |
| RUO | |
| Hamster | |
| This Rabbit anti-Hamster IgG (H+L) Secondary Antibody was developed against purified hamster IgG (H&L). | |
| Secondary | |
| Store at 4°C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title