Learn More
Invitrogen™ Rabbit anti-Human IgE Recombinant Secondary Antibody
Shop All Thermo Scientific ProductsDescription
This antibody reacts to human IgE. No cross reactivity with human IgG, IgM, IgD, or IgA. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Specifications
Specifications
| Antigen | Human IgE |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | RM122 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH 7.2-7.4 |
| Host Species | Rabbit |
| Immunogen | Peptide corresponding to Human IgE. |
| Purification Method | Protein A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.