missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Human IgG, Whole Molecule Polyclonal Secondary Antibody, MP Biomedicals

Product Code. 11493656 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493656 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11493656 Supplier MP Biomedicals Supplier No. 0855008
  • ONLINE EXCLUSIVE PRICE
    261.00 €
    246.75 € / 2mL
    Save 14.25 €
    5% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Host Species: Rabbit
Species Reactivity: Hu

Immunostaining

Specifications

Antigen Human IgG
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Quantity 2 mL
Primary or Secondary Secondary
Target Species Human
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.