missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgG Fc Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-72639
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| IgG Fc | |
| Polyclonal | |
| Alkaline Phosphatase (AP) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| No reaction was observed against Mouse IgG F(ab). | |
| Store at 4C. Do not freeze. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2500, ELISA 1:5000 - 1:15000, Immunohistochemistry 1:200 - 1:1000 | |
| Rabbit anti-Mouse IgG Fc fragment Secondary Antibody was produced by repeated immunization with mouse IgG F(c) fragment in rabbit. | |
| 1 mg | |
| Secondary | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction