missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rabbit anti-Mouse IgG1 Secondary Antibody, HRP

Product Code. 11523771
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11523771 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11523771 Supplier Invitrogen™ Supplier No. PA186329
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Secondary Antibody

PA1-86329 is specific for mouse IgG1 and shows minimal (less than 0.5%) cross-reactivity with mouse IgG2a, IgGb, IgG3 and IgM Reconstitute with 1.0 mL of distilled water. Prior to reconstitution store at 4C. After reconstitution store at -20C. Rabbit anti Mouse IgG1 antibody is specific for mouse IgG1 and shows minimal (<0.5%) cross-reactivity with mouse IgG2a, IgG2b, IgG3 and IgM.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG1
Applications ELISA, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate HRP
Formulation PBS with 1% BSA and 0.01% gentamicin sulfate
Host Species Rabbit
Immunogen Mouse IgG, gamma 1 heavy chain.
Purification Method Affinity chromatography
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 4°C
Product Type Secondary Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.