Learn More
Invitrogen™ Rabbit anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody
Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen™ 31219
Description
Product # 31219 has been successfully used in Western blot, IF, ICC, ELISA and IHC applications. Product # 31219 reacts with the heavy chains of rat IgG and with the light chains common to most rat immunoglobulins, but does not react against non-immunoglobulin serum proteins. This antibody cross-reacts with guinea pig IgG and hamster IgG (1%), and it has minimum cross-reactivity with immunoglobulins from other species.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Rat IgG (H+L) Cross-Adsorbed | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| 500 μg | |
| Secondary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 20mg/mL sucrose and 0.08% sodium azide; pH 7.8 | |
| Affinity chromatography | |
| RUO | |
| Rat | |
| Secondary Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.