missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Sheep IgM (H+L) Secondary Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68363B
This item is not returnable.
View return policy
Description
IgM (H+L) Polyclonal antibody specifically detects IgM (H+L) in Sheep samples. It is validated for ELISA
Specifications
| IgM (H+L) | |
| Polyclonal | |
| PBS | |
| Ovine IgM | |
| 1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| Biotin | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction